Potassium hexacyanocobaltate(III), 90+%
Potassium hexacyanocobaltate(III), 90+%
Potassium hexacyanocobaltate(III), 90+%
Potassium hexacyanocobaltate(III), 90+%
Thermo Scientific Chemicals
Potassium hexacyanocobaltate(III), 90+%
CAS: 13963-58-1
Have Questions?
Catalog NumberQuantity
ALF023126.30250 g
Catalog number ALF023126.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium hexacyanocobaltate(III)
Melting Point(decomposition)
CAS13963-58-1
ColorBrown to Yellow
Health Hazard 1Danger
View more
It is applied in formation of a xerogel via a cyanogel, the gel has potential use in solid-state gas sensors. It is also used as reagent.