Acetonitrile, HPLC Grade, 99.7+% min, Thermo Scientific Chemicals
Acetonitrile, HPLC Grade, 99.7+% min, Thermo Scientific Chemicals
Acetonitrile, HPLC Grade, 99.7+% min, Thermo Scientific Chemicals
Acetonitrile, HPLC Grade, 99.7+% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetonitrile, HPLC Grade, 99.7+% min, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF022927.K21 L
Catalog number ALF022927.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialAcetonitrile
CAS75-05-8
ColorColorless
Health Hazard 1H225-H302-H312-H332-H319: Highly flammable liquid and vapour. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Causes serious eye irritation.
AppearanceColorless liquid
View more
Acetonitrile is used as a mobile phase in high performance liquid chromatography and liquid chromatography coupled with mass spectrometry.