1-Butanol, HPLC Grade, 99%
1-Butanol, HPLC Grade, 99%
1-Butanol, HPLC Grade, 99%
1-Butanol, HPLC Grade, 99%
Thermo Scientific Chemicals
1-Butanol, HPLC Grade, 99%
Filtered through 0.2μm filters | CAS: 71-36-3 | C4H10O | 74.123 g/mol
Have Questions?
Catalog NumberQuantity
ALF022925.K21 L
Catalog number ALF022925.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or Material1-Butanol
Melting Point−90°C to -89°C
CAS71-36-3
Health Hazard 1H226-H302-H315-H318-H335-H336
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P314-P330-P332+P313-P363-P370+P378q-P501c
View more
1-Butanol is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.