Chloroform, HPLC Grade, 99.5+% min
Chloroform, HPLC Grade, 99.5+% min
Chloroform, HPLC Grade, 99.5+% min
Chloroform, HPLC Grade, 99.5+% min
Thermo Scientific Chemicals
Chloroform, HPLC Grade, 99.5+% min
CAS: 67-66-3 | CHCl3 | 119.37 g/mol
Have Questions?
Catalog NumberQuantity
ALF022920.K21 L
Catalog number ALF022920.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialChloroform
Melting Point°C to 63°C
CAS67-66-3
ColorColorless
Health Hazard 1H302-H315-H319-H331-H335-H336-H351-H361d-H373
View more
Chloroform is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.