Ethyl acetate, HPLC Grade, 99.5+%
Ethyl acetate, HPLC Grade, 99.5+%
Ethyl acetate, HPLC Grade, 99.5+%
Ethyl acetate, HPLC Grade, 99.5+%
Thermo Scientific Chemicals
Ethyl acetate, HPLC Grade, 99.5+%
Maximum level of impurities: Evaporation residue 3ppm, Chlorinated pesticides 5ppt, H2O 0.05% | CAS: 141-78-6 | C4H8O2 | 88.106 g/mol
Have Questions?
Catalog NumberQuantity
ALF022912.K21 L
Catalog number ALF022912.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialEthyl acetate
Melting Point°C to 84°C
CAS141-78-6
ColorColorless
Health Hazard 1H225-H319-H336
View more
Ethyl acetate in its highly pure form is used as a solvent for spectrophotometric applications. It is most suited for HPLC instruments and spectrophotometers including UV-visible spectrophotometer, IR spectrometer and mass spectrometer. In addition, it can also be used as a solvent in column chromatography and solvent extraction. Also, it is used in glues, nail polish removers, decaffeinating tea and coffee, and cigarettes. It is present in confectioneries, perfumes, and fruits.