n-Heptane, HPLC grade, 99+%
n-Heptane, HPLC grade, 99+%
n-Heptane, HPLC grade, 99+%
n-Heptane, HPLC grade, 99+%
Thermo Scientific Chemicals
n-Heptane, HPLC grade, 99+%
CAS: 142-82-5 | C7H16 | 100.21 g/mol
Have Questions?
Catalog NumberQuantity
ALF022911.K21 L
Catalog number ALF022911.K2
Price (MYR)
580.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
580.00
EA
Add to cart
Specifications
Chemical Name or Materialn-Heptane
Melting Point91°C
CAS142-82-5
Health Hazard 1H225-H304-H315-H319-H336-H373
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P331-P332+P313-P363-P370+P378q-P501c
View more
n-Heptane is used in paints and coatings, as the rubber cement solvent and GC analysis, HPLC, spectrophotometry and environmental testing. Used as a primary reference standard in determining the knock rating of carburetor fuels and solvent especially for vegetable oils, low temperature thermo- meters, calibrations, and polymerization reaction medium. The hydroisomerization of n-heptane to form dibranched and tribranched products useful for producing high octane gasoline. water/AOT/n-heptane microemulsions plays an essential role for the synthesis, size control, and passivation of CdS nanoparticles.