Methanol, ultrapure, HPLC Grade, 99.8+%
Methanol, ultrapure, HPLC Grade, 99.8+%
Methanol, ultrapure, HPLC Grade, 99.8+%
Methanol, ultrapure, HPLC Grade, 99.8+%
Thermo Scientific Chemicals
Methanol, ultrapure, HPLC Grade, 99.8+%
CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Catalog NumberQuantity
ALF022909.K21 L
Catalog number ALF022909.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialMethanol
Melting Point98°C
CAS67-56-1
Health Hazard 1H225-H301+H311+H331-H370-H372
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P307+P311-P311-P312-P330-P363-P370+P378q-P501c
View more
Methanol is useful in the synthesis of nucleic acids and peptides. It is used as an antifreeze, solvent, fuel, and as a denaturant for ethanol. It is also used for producing biodiesel. It is used for the synthesis of formaldehyde and acetic acid. It is also useful to make chemicals, to remove water from automotive and aviation fuels, as a solvent for paints and plastics, and as an ingredient in a wide variety of products. It plays an important role for the measurement of methyl ester content of pectin. It is also used as a mobile phase in High Performance Liquid Chromatography.