Isobutanol, HPLC Grade, 99+%
Isobutanol, HPLC Grade, 99+%
Isobutanol, HPLC Grade, 99+%
Isobutanol, HPLC Grade, 99+%
Thermo Scientific Chemicals
Isobutanol, HPLC Grade, 99+%
Maximum level of impurities: Evaporation residue 5ppm, H2O 0.05% | CAS: 78-83-1 | C4H10O | 74.123 g/mol
Have Questions?
Catalog NumberQuantity
ALF022908.K21 L
Catalog number ALF022908.K2
Price (MYR)
624.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
624.00
EA
Add to cart
Specifications
Chemical Name or MaterialIsobutanol
CAS78-83-1
Health Hazard 1H226-H315-H318-H335-H336
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P314-P332+P313-P363-P370+P378q-P501c
Melting Point-108°C
View more
Isobutanol is a reagent used in organic reactions. It is employed in the synthesis of new fluorinating reagents. It is used as a mobile phase in High Performance Liquid Chromatography. It is also used in the lipase-catalyzed production of biodiesel as an energy source. It acts as a catalyst for liquid-phase esterification of acetic acid by ion-exchange resins. Dehydration of fermented isobutanol is useful for the production of renewable chemicals and fuels.