Boron trifluoride, 99+%, Thermo Scientific Chemicals
Boron trifluoride, 99+%, Thermo Scientific Chemicals
Boron trifluoride, 99+%, Thermo Scientific Chemicals
Boron trifluoride, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Boron trifluoride, 99+%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF022874.22100 g
Catalog number ALF022874.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Boiling Point-100°C
Chemical Name or MaterialBoron trifluoride
Melting Point-127°C
CAS7637-07-2