Toluene, Semiconductor Grade, 99% min
Toluene, Semiconductor Grade, 99% min
Toluene, Semiconductor Grade, 99% min
Toluene, Semiconductor Grade, 99% min
Thermo Scientific Chemicals
Toluene, Semiconductor Grade, 99% min
Maximum level of impurities: Color (APHA) 10, Acidity 0.2meq/g, Evaporation residue 2ppm, H2O 0.03%, Substances darkened by H2SO4 P.T., Sulfur compounds (as S) 0.003%, Cl 2ppm, PO4 0.5ppm | CAS: 108-88-3 | C7H8 | 92.14 g/mol
Have Questions?
Catalog NumberQuantity
ALF019399.K21 L
Catalog number ALF019399.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialToluene
Melting Point°C to 93°C
CAS108-88-3
ColorColorless
Health Hazard 1H225-H304-H315-H319-H336-H361d-H373
View more
Toluene is used as a solvent in various chemical industries. It is used in the production of various petrochemicals like paint, explosives, urethane foam and phenol formaldehyde used for electrical switches. It is used as a thinner in the paint industry, as a fuel additive to increase octane ratings and as a coolant for its good heat transfer capabilities in sodium cold traps used in nuclear reactor system loops.