Water, ultrapure, Spectrophotometric Grade
Water, ultrapure, Spectrophotometric Grade
Water, ultrapure, Spectrophotometric Grade
Water, ultrapure, Spectrophotometric Grade
Thermo Scientific Chemicals
Water, ultrapure, Spectrophotometric Grade
Maximum level of impurities: Evaporation residue 2ppm, Specific conductance at 25° 1.5μ mhos/cm, SiO2 0.01ppm, Heavy metals (as Pb) 0.01ppm, Substances reducing permanganate P.T. | CAS: 7732-18-5 | H2O | 18.015 g/mol
Have Questions?
Catalog NumberQuantity
ALF019391.K21 L
Catalog number ALF019391.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialWater
CAS7732-18-5
ColorColorless
Melting Point0°C
Boiling Point100°C
View more
Water ultrapure is used in spectrophotometry and environmental testing. It is also used in conductometric titration.