Sodium orthosilicate, (mixture of NaOH and Na2SiO3 yielding ≈ Na4SiO4 in solution), Thermo Scientific Chemicals
Sodium orthosilicate, (mixture of NaOH and Na2SiO3 yielding ≈ Na4SiO4 in solution), Thermo Scientific Chemicals
Sodium orthosilicate, (mixture of NaOH and Na2SiO3 yielding ≈ Na4SiO4 in solution), Thermo Scientific Chemicals
Sodium orthosilicate, (mixture of NaOH and Na2SiO3 yielding ≈ Na4SiO4 in solution), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium orthosilicate, (mixture of NaOH and Na2SiO3 yielding ≈ Na4SiO4 in solution), Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF017568.A11 kg
Catalog number ALF017568.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium orthosilicate
Name NoteMixture of NaOH and Na2SiO3 yielding ≈ Na4SiO4 in solution
CAS13472-30-5
ColorWhite
Health Hazard 1H302-H314-H335
View more
It is used as a degreasant, industrial alkaline detergents, water treatment (boiler-feed); adhesives, builder in soaps.