Sodium 1-Octanesulfonate Monohydrate, 99+%
Sodium 1-Octanesulfonate Monohydrate, 99+%
Sodium 1-Octanesulfonate Monohydrate, 99+%
Sodium 1-Octanesulfonate Monohydrate, 99+%
Thermo Scientific Chemicals
Sodium 1-Octanesulfonate Monohydrate, 99+%
CAS: 207596-29-0 | C8H19NaO4S | 234.29 g/mol
Have Questions?
Catalog NumberQuantity
ALF015764.1850 g
Catalog number ALF015764.18
Price (MYR)
1,496.00
EA
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
1,496.00
EA
Add to cart
Specifications
Chemical Name or MaterialSodium 1-Octanesulfonate Monohydrate
CAS207596-29-0
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Melting Point>300°C
View more
Sodium 1-octanesulfonate monohydrate is used as an anionic detergent. It is an ion pairing reagent used in the HPLC analysis of proteins and peptides. Further, it is used as a surface active agent.