Tin(II) iodide, ultra dry, 99.999% (metals basis)
Tin(II) iodide, ultra dry, 99.999% (metals basis)
Tin(II) iodide, ultra dry, 99.999% (metals basis)
Tin(II) iodide, ultra dry, 99.999% (metals basis)
Thermo Scientific Chemicals
Tin(II) iodide, ultra dry, 99.999% (metals basis)
CAS: 10294-70-9 | I2Sn | 372.519 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALF014476.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Boiling Point720°C (decomposition)
Chemical Name or MaterialTin(II) Iodide
Melting Point320°C
Name NoteAmpouled under argon
CAS10294-70-9
View more
Tin(II) iodide is used to prepare organic-inorganic perovskite thin films which exhibit uniform surfaces and strong photoluminescence. It finds application as an analytical reagent in the determination of rhodium and iridium by spectrophotometry. It is also involved in the synthesis of CH3(CH2)11NH3]SnI3 which is a hybrid semiconductor with MoO3 type tin(II) iodide layers.