Lead(II) oxide, 99.99% (metals basis)
Lead(II) oxide, 99.99% (metals basis)
Lead(II) oxide, 99.99% (metals basis)
Lead(II) oxide, 99.99% (metals basis)
Thermo Scientific Chemicals
Lead(II) oxide, 99.99% (metals basis)
CAS: 1317-36-8 | OPb | 223.20 g/mol
Have Questions?
Catalog NumberQuantity
ALF014240.1850 g
Catalog number ALF014240.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialLead(II) oxide
Melting Point886°C
CAS1317-36-8
ColorYellow
Health Hazard 1H302+H332-H350-H360Df-H372
View more
Lead(II) oxide is employed mostly in lead-based industrial glass and industrial ceramics, including computer components. It is used as an intermediate/precursor in the manufacture of several products, for example water proof cements, lubricants, lubricating oils, inorganic pigments, lead soaps, petroleum refining, rubber, cathode ray tube glass, and polyvinyl chloride (PVC). It is useful for lead acid batteries as cathode and anode. Lead monoxide scores significant applications in oil, gas and chemical manufactures. It is an efficient catalyst for condensation reactions in organic synthesis.