Lanthanum, plasma standard solution, Specpure™ La 1000μg/mL
Lanthanum, plasma standard solution, Specpure™ La 1000μg/mL
Lanthanum, plasma standard solution, Specpure™ La 1000μg/mL
Lanthanum, plasma standard solution, Specpure™ La 1000μg/mL
Thermo Scientific Chemicals
Lanthanum, plasma standard solution, Specpure™ La 1000μg/mL
CAS: 1312-81-8 | La2O3 in 5% HNO3
Have Questions?
Catalog NumberQuantity
ALF013870.AE100 mL
Catalog number ALF013870.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialLanthanum
CAS1312-81-8
Health Hazard 1H290-H314-H335
CAS7697-37-2
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c