Copper, plasma standard solution, Specpure™ Cu 1000μg/mL
Copper, plasma standard solution, Specpure™ Cu 1000μg/mL
Copper, plasma standard solution, Specpure™ Cu 1000μg/mL
Copper, plasma standard solution, Specpure™ Cu 1000μg/mL
Thermo Scientific Chemicals
Copper, plasma standard solution, Specpure™ Cu 1000μg/mL
CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Catalog NumberQuantity
ALF013867.AP500 mL
Catalog number ALF013867.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialCopper
CAS7440-50-8
Health Hazard 1H290-H315-H318
CAS7732-18-5
Health Hazard 3P234-P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362-P390
View more
Copper, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.