Potassium plasma standard solution, Thermo Scientific™, Specpure™
Have Questions?
Catalog Number
Quantity
ALF013866.AD
50 mL
Catalog number ALF013866.AD
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium
CAS7757-79-1
Health Hazard 1H290-H314-H335
CAS7732-18-5
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.