Aluminum, plasma standard solution, Specpure™ Al 1000μg/mL
Aluminum, plasma standard solution, Specpure™ Al 1000μg/mL
Aluminum, plasma standard solution, Specpure™ Al 1000μg/mL
Aluminum, plasma standard solution, Specpure™ Al 1000μg/mL
Thermo Scientific Chemicals
Aluminum, plasma standard solution, Specpure™ Al 1000μg/mL
CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
50 mL
100 mL
Catalog number ALF013856.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialAluminum
CAS7429-90-5
Health Hazard 1H290-H335
CAS7732-18-5
Health Hazard 3P234-P390