Calcium, plasma standard solution, Specpure™ Ca 1000μg/mL
Calcium, plasma standard solution, Specpure™ Ca 1000μg/mL
Calcium, plasma standard solution, Specpure™ Ca 1000μg/mL
Calcium, plasma standard solution, Specpure™ Ca 1000μg/mL
Thermo Scientific Chemicals
Calcium, plasma standard solution, Specpure™ Ca 1000μg/mL
CAS: 471-34-1 | CCaO3 | 100.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALF013852.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialCalcium
CAS471-34-1
Health Hazard 1H290-H314-H332-H335
CAS7732-18-5
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
View more
Calcium, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.