Calcium plasma standard solution, Thermo Scientific™, Specpure™
Calcium plasma standard solution, Thermo Scientific™, Specpure™
Calcium plasma standard solution, Thermo Scientific™, Specpure™
Calcium plasma standard solution, Thermo Scientific™, Specpure™
Thermo Scientific Chemicals
Calcium plasma standard solution, Thermo Scientific™, Specpure™
Have Questions?
Catalog NumberQuantity
ALF013852.AD50 mL
Catalog number ALF013852.AD
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialCalcium
CAS471-34-1
Health Hazard 1H290-H314-H332-H335
CAS7732-18-5
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.