Germanium, plasma standard solution, Specpure™ Ge 1000μg/mL
CAS: 7440-56-4 | Ge | 72.63 g/mol
Have Questions?
Change view
Quantity:
100 mL
50 mL
Catalog number ALF013841.AE
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialGermanium
CAS7440-56-4
Health Hazard 1H290-H315-H318
CAS7664-39-3
Health Hazard 3P234-P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362-P390