Palladium, plasma standard solution, Specpure™, Pd 1000 μg/mL, Thermo Scientific™
Palladium, plasma standard solution, Specpure™, Pd 1000 μg/mL, Thermo Scientific™
Palladium, plasma standard solution, Specpure™, Pd 1000 μg/mL, Thermo Scientific™
Palladium, plasma standard solution, Specpure™, Pd 1000 μg/mL, Thermo Scientific™
Thermo Scientific Chemicals
Palladium, plasma standard solution, Specpure™, Pd 1000 μg/mL, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALF013833.AE100 mL
Catalog number ALF013833.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialPalladium, plasma standard solution, Specpure™
Health Hazard 1H290-H314-H335
Health Hazard 2GHS H Statement
H314-H335;
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
Recommended StorageAmbient temperatures
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.