Bismuth, plasma standard solution, Specpure™ Bi 1000μg/mL
Bismuth, plasma standard solution, Specpure™ Bi 1000μg/mL
Bismuth, plasma standard solution, Specpure™ Bi 1000μg/mL
Bismuth, plasma standard solution, Specpure™ Bi 1000μg/mL
Thermo Scientific Chemicals
Bismuth, plasma standard solution, Specpure™ Bi 1000μg/mL
CAS: 7440-69-9 | Bi | 208.98 g/mol
Have Questions?
Catalog NumberQuantity
ALF013822.AP500 mL
Catalog number ALF013822.AP
Price (MYR)
740.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
740.00
EA
Add to cart
Specifications
Chemical Name or MaterialBismuth
CAS7440-69-9
Health Hazard 1H290-H314-H335
CAS7732-18-5
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c