Cadmium, plasma standard solution, Specpure™ Cd 1000μg/mL
Cadmium, plasma standard solution, Specpure™ Cd 1000μg/mL
Cadmium, plasma standard solution, Specpure™ Cd 1000μg/mL
Cadmium, plasma standard solution, Specpure™ Cd 1000μg/mL
Thermo Scientific Chemicals
Cadmium, plasma standard solution, Specpure™ Cd 1000μg/mL
CAS: 7440-43-9 | Cd | 112.41 g/mol
Have Questions?
Catalog NumberQuantity
ALF013813.AE100 mL
Catalog number ALF013813.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialCadmium
CAS7440-43-9
Health Hazard 1H290-H315-H318-H350
Health Hazard 2When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
CAS7732-18-5