Nickel(II) formate dihydrate
Nickel(II) formate dihydrate
Thermo Scientific Chemicals
Nickel(II) formate dihydrate
For Nickel Catalyst Preparations | CAS: 15694-70-9 | Ni(HCO2)2·2H2O
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF013801.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialNickel(II) formate dihydrate
Melting Point130°C to 140°C-2H2O
CAS15694-70-9
ColorGreen
Health Hazard 1H317-H334-H341-H350-H360-H372
View more
Nickel(II) formate dihydrate is used in nickel catalyst preparations. It is also used as a biochemical for proteomics research.