Indium(III) 2,4-pentanedionate, 98%
Indium(III) 2,4-pentanedionate, 98%
Indium(III) 2,4-pentanedionate, 98%
Indium(III) 2,4-pentanedionate, 98%
Thermo Scientific Chemicals
Indium(III) 2,4-pentanedionate, 98%
Used in catalysts | CAS: 14405-45-9 | C15H21InO6 | 412.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALF013794.14
Price (MYR)
2,740.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
2,740.00
EA
Add to cart
Specifications
Chemical Name or MaterialIndium(III) 2,4-pentanedionate
CAS14405-45-9
Health Hazard 1H302+H312+H332
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
Melting Point186°C (decomposition)
View more
Used in catalysts.Indium(III) 2,4-pentanedionate is used as a precursor for the preparation of indium(III)oxide. It acts as a catalyst in the synthesis of oxindole derivatives. It finds application in gas sensors. Further, it is used in the synthesis of indium tin oxide nanocrystals. In addition to this, it is involved in the preparation of thin-film solar cells by atomic layer chemical vapor deposition with hydrogen sulfide.