Ammonium iron(II) sulfate hexahydrate, ACS, 98.5-101.5%
Ammonium iron(II) sulfate hexahydrate, ACS, 98.5-101.5%
Ammonium iron(II) sulfate hexahydrate, ACS, 98.5-101.5%
Ammonium iron(II) sulfate hexahydrate, ACS, 98.5-101.5%
Ammonium iron(II) sulfate hexahydrate, ACS, 98.5-101.5%
Thermo Scientific Chemicals
Ammonium iron(II) sulfate hexahydrate, ACS, 98.5-101.5%
Air and Light Sensitive | CAS: 7783-85-9 | FeH20N2O14S2 | 392.13 g/mol
Have Questions?
Catalog NumberQuantity
ALF013448.A32 kg
Catalog number ALF013448.A3
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium iron(II) sulfate hexahydrate
Melting Point∽100°C (decomposition)
Name NoteFor ACS analysis
CAS7783-85-9
ColorBlue-Green
View more
Polymerization catalyst, photographyAmmonium iron(II) sulfate hexahydrate is widely utilized in analytical chemistry for titration purposes since it is affected lesser by oxygen in the air than iron(II) sulpahte. It is employed to measure high doses of gamma rays by using Fricke's dosemeter. It is involved in a variety of other applications from nanomaterials to general redox reactions. It is used in the medical field as an antianemic agent for treatment of iron deficiency.