Check Order Status
Boc-His Tos , Advanced ChemTech
Element Search
Structure Search
Thermo Scientific Chemicals
Boc-His Tos , Advanced ChemTech
Have Questions?
Catalog Number
Quantity
ALF013411.30
5 g
Catalog number
ALF013411.30
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material
Boc-His Tos
CAS
35899-43-5
Color
White to Off-White
Molecular Formula
C18H23N3O6S
InChI Key
DCLJSEPKYJSEHW-UHFFFAOYNA-N
View more