Check Order Status
Boc-Ile.1/2H2O, Advanced ChemTech
Element Search
Structure Search
Thermo Scientific Chemicals
Boc-Ile.1/2H2O, Advanced ChemTech
Have Questions?
Catalog Number
Quantity
ALF013411.18
100 g
Catalog number
ALF013411.18
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material
Boc-Ile.1/2H2O
CAS
13139-16-7
Color
White to Off-White
Molecular Formula
C11H21NO4
MDL Number
MFCD00038324 MFCD00237550
View more