Check Order Status
Fmoc-N-Me-Leu-OH, Advanced ChemTech
Element Search
Structure Search
Thermo Scientific Chemicals
Fmoc-N-Me-Leu-OH, Advanced ChemTech
Have Questions?
Catalog Number
Quantity
ALF013402.06
5 g
Catalog number
ALF013402.06
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material
Fmoc-N-Me-Leu-OH
CAS
103478-62-2
Color
White/Off-White
Molecular Formula
C22H25NO4
MDL Number
MFCD00151933
View more