Check Order Status
Fmoc-Cit-OH, Advanced ChemTech
Element Search
Structure Search
Thermo Scientific Chemicals
Fmoc-Cit-OH, Advanced ChemTech
Have Questions?
Catalog Number
Quantity
ALF013398.14
25 g
Catalog number
ALF013398.14
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material
Fmoc-Cit-OH
CAS
133174-15-9
Color
White/Off-White
Molecular Formula
C21H23N3O5
InChI Key
NBMSMZSRTIOFOK-UHFFFAOYNA-N
View more