Gadolinium(III) nitrate hydrate, 99.5% (REO)
Gadolinium(III) nitrate hydrate, 99.5% (REO)
Gadolinium(III) nitrate hydrate, 99.5% (REO)
Gadolinium(III) nitrate hydrate, 99.5% (REO)
Gadolinium(III) nitrate hydrate, 99.5% (REO)
Thermo Scientific Chemicals
Gadolinium(III) nitrate hydrate, 99.5% (REO)
CAS: 94219-55-3 | GdN3O9 | 343.26 g/mol
Have Questions?
Catalog NumberQuantity
ALF012917.1425 g
Catalog number ALF012917.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialGadolinium(III) nitrate hydrate
Melting Point92°C
CAS94219-55-3
ColorWhite
Health Hazard 1H272-H315-H319-H335
View more
It is applied as an oxidizing agent and the precursor material in the preparation of gadolinium compounds. It is extensively employed in the production of specialized glasses and ceramics. It is also employed in heavy water nuclear reactors.