Ruthenium(III) nitrosylnitrate solution, Ru 1.5% w/v
Ruthenium(III) nitrosylnitrate solution, Ru 1.5% w/v
Ruthenium(III) nitrosylnitrate solution, Ru 1.5% w/v
Ruthenium(III) nitrosylnitrate solution, Ru 1.5% w/v
Thermo Scientific Chemicals
Ruthenium(III) nitrosylnitrate solution, Ru 1.5% w/v
CAS: 34513-98-9
Have Questions?
Catalog NumberQuantity
ALF012630.AD50 mL
Catalog number ALF012630.AD
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialRuthenium(III) nitrosylnitrate solution
ColorRed to Brown
CAS34513-98-9
Health Hazard 1H290-H314-H332
Health Hazard 2GHS H Statement
H272-H314
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.
View more
Ruthenium(III) nitrosyl nitrate used as a catalyst in various reactions namely hydrogenation, carbonylation, hydrosilylation, hydrofomylation, C-X coupling (eg Heck, Suzuki, Stille) and chiral catalysis reactions. It is also used in the production of thick-film resistors.