Chromium(VI) oxide, 99%
Chromium(VI) oxide, 99%
Chromium(VI) oxide, 99%
Chromium(VI) oxide, 99%
Thermo Scientific Chemicals
Chromium(VI) oxide, 99%
CAS: 1333-82-0 | CrO3 | 99.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number ALF012522.A1
Price (MYR)
560.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
560.00
EA
Add to cart
Specifications
Boiling Point250°C (decomposition)
Chemical Name or MaterialChromium(VI) oxide
Melting Point196°C
CAS1333-82-0
Health Hazard 1H271-H301-H310+H330-H314-H317-H334-H335-H340-H350-H361f-H372
View more
It is widely used in chromium electroplating, and as protective coatings in corrosion/oxidation resistance of metals. It is also employed in CDs and DVDs. It is useful as an oxidant in Jones oxidation in acetic acid or acetone. It is used in the oxidation of primary alcohols into carboxylic acids, and secondary alcohols into ketones. With phosphoric acid, it is used as a stripping agent for anodic coatings of all types and in the production of syntheric rubies.