Nickel(II) sulfate hexahydrate, 98%
Nickel(II) sulfate hexahydrate, 98%
Nickel(II) sulfate hexahydrate, 98%
Nickel(II) sulfate hexahydrate, 98%
Nickel(II) sulfate hexahydrate, 98%
Thermo Scientific Chemicals
Nickel(II) sulfate hexahydrate, 98%
In nickel plating, as mordant in dyeing and in printing textiles | CAS: 10101-97-0 | H12NiO10S | 262.839 g/mol
Have Questions?
Catalog NumberQuantity
ALF012514.36500 g
Catalog number ALF012514.36
Price (MYR)
517.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
517.00
EA
Add to cart
Specifications
Chemical Name or MaterialNickel(II) sulfate hexahydrate
CAS10101-97-0
Health Hazard 1H302-H315-H317-H319-H332-H334-H335-H341-H350i-H360D-H372
Health Hazard 2GHS H Statement
H334-H350-H360-H372-H341-H302-H332-H315-H317
Health Hazard 3P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P342+P311-P362-P501c
View more
Nickel sulfate hexahydrate is mainly used in Ni plating, as mordant in dyeing and in printing textiles, as a blackening agent for zinc and brass and in the fabrication of organic nickel salts. It is used as an effective catalyst for gas-phase dehydration of glycerol to acrolein. Nickel sulfate hexahydrate crystal is an efficient ultraviolet filter material. Columns used in biochemistry and molecular biology can be regenerated with nickel sulfate. It is used to prepare nickel carbonate, a precursor to pigments and nickel-based catalysts.