Triethylamine, 99+%
Triethylamine, 99+%
Triethylamine, 99+%
Thermo Scientific Chemicals
Triethylamine, 99+%
CAS: 121-44-8 | C6H15N | 101.193 g/mol
Have Questions?
Catalog NumberQuantity
ALF012391.30250 g
Catalog number ALF012391.30
Price (MYR)
390.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
390.00
EA
Add to cart
Specifications
Chemical Name or MaterialTriethylamine
CAS121-44-8
Health Hazard 1H225-H302-H311+H331-H314-H335-H336
Health Hazard 2GHS H Statement
H225-H314-H302-H312-H332
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
View more
Triethylamine is a base used to prepare esters and amides from acyl chlorides as well as in the synthesis of quaternary ammonium compounds. It acts as a catalyst in the formation of urethane foams and epoxy resins, dehydrohalogeantion reactions, acid neutralizers for condensation reactions and Swern oxidations. It finds application in reverse phase high-performance liquid chromatography (HPLC) as a mobile-phase modifier. It is also used as an accelerator activator for rubber, as a propellant, as a corrosion inhibitor, as a curing and hardening agent for polymers and for the desalination of seawater. Furthermore, it is used in the automotive casting industry and the textile industry.