Magnesium hydroxide, 95-100.5%
Magnesium hydroxide, 95-100.5%
Magnesium hydroxide, 95-100.5%
Magnesium hydroxide, 95-100.5%
Thermo Scientific Chemicals
Magnesium hydroxide, 95-100.5%
Flame Retardant in Plastics, Such As PVC, As It Produces Less Toxic Fumes | CAS: 1309-42-8 | H2MgO2 | 58.32 g/mol
Have Questions?
Catalog NumberQuantity
ALF012367.36500 g
Catalog number ALF012367.36
Price (MYR)
459.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
459.00
EA
Add to cart
Specifications
Chemical Name or MaterialMagnesium hydroxide
Melting Point360°dec
CAS1309-42-8
ColorWhite
Health Hazard 1H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
View more
It is employed as an additive in certain refractories. It is applied as a non-hazardous alkali in neutralizing acidic wastewater. It is also used in making artificial reefs. It widely used in milk of magnesia antacids and laxatives. Also works as an antiperspirant.