Barium chloride dihydrate, ACS
Barium chloride dihydrate, ACS
Barium chloride dihydrate, ACS
Barium chloride dihydrate, ACS
Thermo Scientific Chemicals
Barium chloride dihydrate, ACS
CAS: 10326-27-9 | BaCl2H4O2 | 244.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
500 g
Catalog number ALF012310.A3
Price (MYR)
1,145.00
EA
Add to cart
Quantity:
2 kg
Request bulk or custom format
Price (MYR)
1,145.00
EA
Add to cart
Specifications
Chemical Name or MaterialBarium chloride dihydrate
Melting Point113°C (in -2H2O)
Name Notedihydrate
CAS10326-27-9
Health Hazard 1H301-H319-H332
View more
Barium chloride is usually employed as a chemical reagent to precipitate soluble sulfates and also in the purification of the seawater solution. It is practiced in several applications like heat-treating baths, waste water treatment, the production of PVC stabilizers, oil lubricants, fireworks materials, hardening of steel, in the manufacture of pigments and in the manufacture of other barium salts. It is applied to prepare composites to absorb ammonia and to increase the crystallinty of polymers.