Magnesium chloride hexahydrate, 97+%
Magnesium chloride hexahydrate, 97+%
Magnesium chloride hexahydrate, 97+%
Magnesium chloride hexahydrate, 97+%
Thermo Scientific Chemicals
Magnesium chloride hexahydrate, 97+%
CAS: 7791-18-6 | Cl2H12MgO6 | 203.30 g/mol
Have Questions?
Catalog NumberQuantity
ALF012288.36500 g
Catalog number ALF012288.36
Price (MYR)
426.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
426.00
EA
Add to cart
Specifications
Chemical Name or MaterialMagnesium chloride hexahydrate
Melting Point117°C (decomposition)
CAS7791-18-6
ColorWhite
Recommended StorageAmbient temperatures
View more
It is utilized in the polymerase chain reactions (PCR). It is used for dust control, ice control, and road stabilization. It is employed in a variety of applications such as fertilizers, mineral supplements for animals, waste water treatment, artificial seawater, textiles, paper, fireproofing agents, cements, and fire extinguishers. Pharmaceutical preparations are used in several medicines and topical or skin related applications. It acts as a coagulant in the preparation of tofu from soy milk.