Strontium chloride, anhydrous, 95%
Strontium chloride, anhydrous, 95%
Strontium chloride, anhydrous, 95%
Strontium chloride, anhydrous, 95%
Thermo Scientific Chemicals
Strontium chloride, anhydrous, 95%
CAS: 10476-85-4
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
500 g
Catalog number ALF012202.A3
Price (MYR)
1,402.00
EA
Add to cart
Quantity:
2 kg
Request bulk or custom format
Price (MYR)
1,402.00
EA
Add to cart
Specifications
Boiling Point1250°C
Chemical Name or MaterialStrontium chloride
Melting Point873°C
Name NoteAnhydrous
CAS10476-85-4
View more
It is widely used in fireworks for the source of red color. It is also used in dental and biological research. It is the precursor material to other compounds of strontium. It is used to prepare yellow strontium chromate, a corrosion inhibitor for aluminum. It is also employed in glass-making, sea water aquaria, and metallurgy.