Ruthenium powder, -200 mesh, 99.95% (metals basis)
Ruthenium powder, -200 mesh, 99.95% (metals basis)
Ruthenium powder, -200 mesh, 99.95% (metals basis)
Ruthenium powder, -200 mesh, 99.95% (metals basis)
Thermo Scientific Chemicals
Ruthenium powder, -200 mesh, 99.95% (metals basis)
CAS: 7440-18-8 | Ru | 101.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
Catalog number ALF012061.09
Price (MYR)
7,488.00
EA
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (MYR)
7,488.00
EA
Add to cart
Specifications
Chemical Name or MaterialRuthenium powder
CAS7440-18-8
Health Hazard 1H228-H500
Health Hazard 2GHS H Statement
H228
Flammable solid.
Health Hazard 3P210-P240-P241-P280-P370+P378q