Zinc foil, 0.1mm (0.004in) thick, Puratronic™, 99.994% (metals basis)
Zinc foil, 0.1mm (0.004in) thick, Puratronic™, 99.994% (metals basis)
Zinc foil, 0.1mm (0.004in) thick, Puratronic™, 99.994% (metals basis)
Zinc foil, 0.1mm (0.004in) thick, Puratronic™, 99.994% (metals basis)
Thermo Scientific Chemicals
Zinc foil, 0.1mm (0.004in) thick, Puratronic™, 99.994% (metals basis)
Form: approx. 4.5g/50 x 50mm | CAS: 7440-66-6 | Zn | 65.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 x 300 mm
50 x 50 mm
100 x 100 mm
Catalog number ALF011912.HG
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 x 300 mm
Request bulk or custom format
Specifications
Chemical Name or MaterialZinc
ColorGray
CAS7440-66-6
AppearanceGray foil