SiliaSep™ Flash Cartridges, Silica-Based, Cyano (CN), 40 to 63 μm, 60 Å
SiliaSep™ Flash Cartridges, Silica-Based, Cyano (CN), 40 to 63 μm, 60 Å
Actual product may vary
SiliaSep™ Flash Cartridges, Silica-Based, Cyano (CN), 40 to 63 μm, 60 Å
SiliaSep™ Flash Cartridges, Silica-Based, Cyano (CN), 40 to 63 μm, 60 Å
Thermo Scientific Chemicals
SiliaSep™ Flash Cartridges, Silica-Based, Cyano (CN), 40 to 63 μm, 60 Å
SiliaSep™ Flash Cartridges, Silica-Based, Cyano (CN), 40 - 63 μm, 60 Å (4 g to 1600 g) are packed with Cyano sorbent for purification of polar organic compounds in reversed-phase or normal phase.
Have Questions?
Catalog NumberColumn TypeMax. PressureDiameter (Metric) OuterQuantity
ALF011837.2212 g Flash Cartridge225 psi/16 bar21 mm1 Pk.
Catalog number ALF011837.22
Price (MYR)
546.00
EA
Add to cart
Column Type:
12 g Flash Cartridge
Max. Pressure:
225 psi/16 bar
Diameter (Metric) Outer:
21 mm
Quantity:
1 Pk.
Price (MYR)
546.00
EA
Add to cart
  • Directly compatible with a variety of flash systems (slip tip connection type, but can be used with many luer lock systems)
  • Pre-packed disposable flash cartridges
  • Unique one-piece polypropylene cartridge for high pressure and solvent resistance
  • Leak-free guaranteed
  • Made with high quality silica (40 - 63 μm, 60 Å)
  • Tight particle size distribution
  • Low fines
  • Neutral pH
  • Controlled water content
  • Very diversified formats (4, 12, 25, 40, 80, 120, 220, 330, 800, and 1600 g)
  • Effective packing technology
  • Good resolution, no tailing
  • Stringent quality control (ISO 9001:2015 Registered)
  • Lot-to-lot reproducibility
  • For 800 g and 1600 g XL formats, an adapter may be needed to connect the cartridge to the system
Specifications
Column Type12 g Flash Cartridge
Diameter (Metric) Outer21 mm
For Use With (Application)Purification of polar organic compounds in reversed-phase or normal phase
Length (Metric)117 mm
MaterialPolypropylene
Max. Pressure225 psi/16 bar
TypeFlash Cartridge
Capacity (Metric)0.060 to 0.600 g
Carbon Load7%
Particle Size40 to 63 μm
EndcappedYes
For Use With (Equipment)Direct compatibility with most flash systems of the market
Particle ShapeIrregular
pH2 to 10
Pore Size60 Å
Product LineSiliaSep
Quantity1 Pk.
Stationary PhaseCyano (2-Cyano, CN, Nitrile)
Surface Area (Metric)500 m2/g
Unit SizeEA
Contents & Storage
Keep dry.