R 1405 Propeller
R 1405 Propeller
Actual product may vary
Thermo Scientific Chemicals
R 1405 Propeller
The R 1405 propeller head is designed for the use with the overhead stirrers EUROSTAR 20 high speed digital/control and T 50 digital.
Have Questions?
Catalog number ALF011832.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
  • It is ideally suited for applications which work with low to medium mixing viscosities that require axial flow pattern at high speed with minimal temperature increase
  • Working range: 0.25 to 30 L
  • Rotor diameter: 45 mm
  • Note: R 1405 needs to be attached to either the precision shaft R 6000 (if used with EUROSTAR 20 high speed) or the high speed setting shaft R 50 (if used with T 50 digital) in order to operate. The shafts are not included in the scope of delivery of R 1405.
Specifications
Diameter (Metric) Fan45 mm
For Use With (Equipment)Overhead Stirrers EUROSTAR 20
Weight (Metric)0.0261 kg
Stirring Range0.25 to 30 L
Unit SizeEA