SiliaPrepX™ SPE Cartridges - HLB
SiliaPrepX™ SPE Cartridges - HLB
Actual product may vary
Thermo Scientific Chemicals
SiliaPrepX™ SPE Cartridges - HLB
SPE Cartridges packed with wettable copolymer presenting a Hydrophilic-Lipophilic Balance (HLB), for extraction of neutral, acidic or basic compounds.
Have Questions?
Catalog NumberBed WeightVolume (Metric)Quantity
ALF011829.3030 mg3 mL50 Pk.
Catalog number ALF011829.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Bed Weight:
30 mg
Volume (Metric):
3 mL
Quantity:
50 Pk.
  • For use with vacuum manifolds and automated SPE equipments
  • Tightly packed and homogeneous silica bed for reliability and increased productivity
  • Tight particle size distribution and very good packing with no fines, allowing excellent separation
  • High recoveries and yields
  • Reproducible flow rates lot-to-lot
  • Flanged polypropylene tubes
  • 20 μm polyethylene frits
Specifications
Adsorbent TypePolymer
Carbon Load0.88
Bed Weight30 mg
For Use With (Application)Extraction of drugs and metabolites from biological fluids, API from tablets and cream
For Use With (Equipment)Vacuum Manifolds or Automated SPE Equipment
Particle Size40 μm
Product LineSiliaPrepX
SorbentHLB (Hydrophilic-Lipophilic Balance)
TypeSPE Cartridge
Volume (Metric)3 mL
MaterialPolypropylene (PP) tube, Polyethylene (PE) frits
Particle ShapeSpherical
Pore Size110 Å
Quantity50 Pk.
Surface Area (Metric)850 m2/g
Tube Size8.9 x 74.7 mm
Length (Metric)74.7 mm
Diameter (Metric) Inner8.9 mm
Unit SizeEA