SiliaPrep™ SPE Cartridges - Florisil LP
SiliaPrep™ SPE Cartridges - Florisil LP
Actual product may vary
Thermo Scientific Chemicals
SiliaPrep™ SPE Cartridges - Florisil LP
SPE Cartridges packed with Florisil 75 to 150 μm, for extraction of moderately polar compounds such as PCBs and polysaccharides.
Have Questions?
Catalog NumberBed WeightVolume (Metric)Quantity
ALF011828.362 g12 mL20 Pk.
Catalog number ALF011828.36
Price (MYR)
1,550.00
EA
Add to cart
Bed Weight:
2 g
Volume (Metric):
12 mL
Quantity:
20 Pk.
Price (MYR)
1,550.00
EA
Add to cart
  • For use with vacuum manifolds and automated SPE equipments
  • Tightly packed and homogeneous silica bed for reliability and increased productivity
  • Tight particle size distribution and very good packing with no fines, allowing excellent separation
  • High recoveries and yields
  • Reproducible flow rates lot-to-lot
  • Flanged polypropylene tubes
  • 20 μm polyethylene frits
Specifications
Adsorbent TypeFlorisil
Bed Weight2 g
For Use With (Application)Extraction of chlorinated pesticides, PCBs and polysaccharides
For Use With (Equipment)Vacuum Manifolds or Automated SPE Equipment
Particle Size75 to 150 μm
Product LineSiliaPrep
SorbentFlorisil (Magnesium Silicate) LP
TypeSPE Cartridge
Volume (Metric)12 mL
MaterialPolypropylene (PP) tube, Polyethylene (PE) frits
Particle ShapeIrregular
Pore Size100 Å
Quantity20 Pk.
Surface Area (Metric)250 m2/g
Tube Size15.8 x 90 mm
Length (Metric)90 mm
Diameter (Metric) Inner15.8 mm
Unit SizeEA