SiliaPrep™ SPE Cartridges - Florisil
SiliaPrep™ SPE Cartridges - Florisil
Actual product may vary
Thermo Scientific Chemicals
SiliaPrep™ SPE Cartridges - Florisil
SPE Cartridges packed with Florisil 40 to 75 μm (TLC grade), for extraction of moderately polar compounds such as PCBs and polysaccharides.
Have Questions?
Catalog NumberBed WeightVolume (Metric)Quantity
ALF011828.22500 mg6 mL50 Pk.
Catalog number ALF011828.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Bed Weight:
500 mg
Volume (Metric):
6 mL
Quantity:
50 Pk.
  • For use with vacuum manifolds and automated SPE equipments
  • Tightly packed and homogeneous silica bed for reliability and increased productivity
  • Tight particle size distribution and very good packing with no fines, allowing excellent separation
  • High recoveries and yields
  • Reproducible flow rates lot-to-lot
  • Flanged polypropylene tubes
  • 20 μm polyethylene frits
Specifications
Adsorbent TypeFlorisil
Bed Weight500 mg
For Use With (Application)Extraction of chlorinated pesticides, PCBs and polysaccharides
For Use With (Equipment)Vacuum Manifolds or Automated SPE Equipment
Particle Size40 to 75 μm
Product LineSiliaPrep
SorbentFlorisil (Magnesium Silicate)
TypeSPE Cartridge
Volume (Metric)6 mL
MaterialPolypropylene (PP) tube, Polyethylene (PE) frits
Particle ShapeIrregular
Pore Size100 Å
Quantity50 Pk.
Surface Area (Metric)250 m2/g
Tube Size12.7 x 77.8 mm
Length (Metric)77.8 mm
Diameter (Metric) Inner12.7 mm
Unit SizeEA