SiliaSep OT Flash Cartridges, Tosic Acid (SCX)
SiliaSep OT Flash Cartridges, Tosic Acid (SCX)
Actual product may vary
Thermo Scientific Chemicals
SiliaSep OT Flash Cartridges, Tosic Acid (SCX)
Open-top Flash Cartridges packed with Tosic Acid sorbent (SCX) for purification of weak basic molecules.
Have Questions?
Catalog NumberBed WeightDiameter (Metric) InnerLength (Metric)Volume (Metric)
ALF011827.3015 g26.8 mm154 mm70 mL
Catalog number ALF011827.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Bed Weight:
15 g
Diameter (Metric) Inner:
26.8 mm
Length (Metric):
154 mm
Volume (Metric):
70 mL
SiliaSep OT Flash Cartridges are for use with vacuum manifolds and automated SPE equipments.
  • Also compatible with FlashMaster™ systems
  • Tightly packed and homogeneous silica bed for reliability and increased productivity
  • Tight particle size distribution and very good packing with no fines, allowing for excellent separation
  • High recoveries and yields
  • Reproducible flow rates lot-to-lot
  • Flanged polypropylene tubes
  • 20 μm polyethylene frits
  • Maximum operating pressure: 60 psi
Specifications
Adsorbent TypeSilica
Carbon Load9%
Column TypeOpen-top SPE Cartridge
Bed Weight15 g
For Use With (Application)Extraction of weak basic molecules, catch and release of amines
For Use With (Equipment)Vacuum Manifolds or Automated SPE Equipment
Particle Size40 to 63 μm
Product LineSiliaSep OT
SorbentTosic Acid (Ethyltosic Acid, SCX)
TypeSPE Cartridge
Volume (Metric)70 mL
EndcappedYes
MaterialPolypropylene tube, Polyethylene frits
Particle ShapeIrregular
Pore Size60 Å
Quantity16 Pk.
Surface Area (Metric)500 m2/g
Length (Metric)154 mm
Diameter (Metric) Inner26.8 mm
Unit SizeEA