Sodium dihydrogen phosphate monohydrate, ACS, 98.0-102.0%
Sodium dihydrogen phosphate monohydrate, ACS, 98.0-102.0%
Thermo Scientific Chemicals
Sodium dihydrogen phosphate monohydrate, ACS, 98.0-102.0%
CAS: 10049-21-5 | H2NaO4P x H2O | 137.99 g/mol
Have Questions?
Catalog NumberQuantity
ALF011591.30250 g
Catalog number ALF011591.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium dihydrogen phosphate monohydrate
Melting Point100°C (in 2H2O)
CAS10049-21-5
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319-H335;
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Sodium phosphate monobasic monohydrate is used as sequestrant, emulsifier, mordant in dyeing, reagent and buffer in foods and analytical chemistry. It is applied as a fireproofing agent and for weighting silk in tanning. It is employed in manufacturing of enamels, ceramics, detergents, boiler compounds, in soldering and brazing instead of borax.